CAS RN 3483-82-7
IUPAC Nombre:
ethyl 3-(4-hydroxyphenyl)-2-(phenylformamido)propanoate
Sinónimos:
ethyl 3-(4-hydroxyphenyl)-2-(phenylformamido)propanoate
Peso molecular (g/mol):
313.35
Fórmula molecular:
C18H19NO4
InChi Key:
SRLROPAFMUDDRC-UHFFFAOYNA-N
SMILES:
CCOC(=O)C(CC1=CC=C(O)C=C1)NC(=O)C1=CC=CC=C1
1
–
3
de
3
resultados
1
Éster de etilo de N-benzoílo-L-tirosina, 98 %, Thermo Scientific Chemicals
CAS: 3483-82-7 Fórmula molecular: C18H19NO4 Peso molecular (g/mol): 313.35 Número MDL: MFCD00002388 Clave InChI: SRLROPAFMUDDRC-XISACWJONA-N Sinónimo: n-benzoyl-l-tyrosine ethyl ester,bz-tyr-oet,ethyl n-benzoyl-l-tyrosinate,btee,ethyl benzoyltyrosinate,benzoyltyrosine ethyl ester,benzoyl-l-tyrosine ethyl ester,s-ethyl 2-benzamido-3-4-hydroxyphenyl propanoate,benzoyl-tyr-oet,l-tyrosine, n-benzoyl-, ethyl ester PubChem CID: 77033 Nombre IUPAC: (2S)-2-benzamido-3-(4-hidroxifenil)propanoato de etilo SMILES: CCOC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)C1=CC=CC=C1