CAS RN 328232-95-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
MK-0557, MedChemExpress
MedChemExpress MK-0557 is a highly selective, orally available neuropeptide Y5 receptor antagonist with a Ki of 1.6 nM.