CAS RN 30271-38-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
24-Hidroxicolesterol, TRC
CAS: 30271-38-6 Fórmula molecular: C27H46O2 Peso molecular (g/mol): 402.65 Sinónimo: Cholest-5-ene-3β,24-diol,(3β)-Cholest-5-ene-3,24-diol Nombre IUPAC: (3S,8S,9S,10R,13R,14S,17R)-17-[(2R)-5-hidroxi-6-metilheptano-2-il]-10,13-dimetil-2,3,4,7,8,9,11,12,14,15,16,17-dodecahidro-1H-ciclopenta[a]fenantotra-3-ol SMILES: CC(C)C(O)CC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
2
24-Hydroxycholesterol, MedChemExpress
MedChemExpress 24-Hydroxycholesterol is a natural sterol, which serves as a positive allosteric modulator of N-Methyl-d-Aspartate (NMDA) receptorsR, and a potent activator of the transcription factors LXR.