CAS RN 300353-13-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
RO-28-1675, MedChemExpress
MedChemExpress RO-28-1675 is a potent allosteric glucokinase (GK) activator with an EC50 of 54 nM. RO-28-1675 can be used for the research of type 2 diabetes.