CAS RN 288150-92-5
IUPAC Nombre:
3-(6,8-difluoro-2-methylquinolin-4-yl)-1-[4-(dimethylamino)phenyl]urea
Sinónimos:
3-(6,8-difluoro-2-methylquinolin-4-yl)-1-[4-(dimethylamino)phenyl]urea
Peso molecular (g/mol):
356.38
Fórmula molecular:
C19H18F2N4O
InChi Key:
JTARFZSNUAGHRB-UHFFFAOYSA-N
SMILES:
CN(C)C1=CC=C(NC(=O)NC2=C3C=C(F)C=C(F)C3=NC(C)=C2)C=C1
1
–
2
de
2
resultados
2
SB-408124, MedChemExpress
MedChemExpress SB-408124 is a non-peptide OX1 receptor antagonist with Kis of 57 nM and 27 nM in whole cell and membrane, respectively. SB-408124 exhibits 50-fold selectivity over OX2 receptor.