CAS RN 245075-84-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
LR-90, MedChemExpress
MedChemExpress LR-90 is an advanced glycation end product (AGE) inhibitor, inhibits inflammatory responses in human monocytes. LR-90 is also used in the research of diabetic animal model.