CAS RN 23109-05-9
IUPAC Nombre:
2-[34-(butan-2-yl)-13-(3,4-dihydroxybutan-2-yl)-8,22-dihydroxy-2,5,11,14,27,30,33,36,39-nonaoxo-27λ⁴-thia-3,6,12,15,25,29,32,35,38-nonaazapentacyclo[14.12.11.0⁶,¹⁰.0¹⁸,²⁶.0¹⁹,²⁴]nonatriaconta-18(26),19,21,23-tetraen-4-yl]acetamide
Sinónimos:
α-amanitin
Peso molecular (g/mol):
918.98
Fórmula molecular:
C39H54N10O14S
InChi Key:
CIORWBWIBBPXCG-UHFFFAOYNA-N
SMILES:
CCC(C)C1NC(=O)CNC(=O)C2CC3=C(NC4=CC(O)=CC=C34)S(=O)CC(NC(=O)CNC1=O)C(=O)NC(CC(N)=O)C(=O)N1CC(O)CC1C(=O)NC(C(C)C(O)CO)C(=O)N2
1
–
2
de
2
resultados
1
Tocris Bioscience™ α-Amanitin
Inhibitor of RNA polymerase II
2
α-Amanitin, MedChemExpress
MedChemExpress α-Amanitin is the principal toxin of several deadly poisonous mushrooms, exerting its toxic function by inhibiting RNA-polymerase II.