CAS RN 224177-60-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
Tocris Bioscience™ Siramesine Hydrochloride
High affinity σ receptor agonist (Ki values are 0.19 and 17 nM for σ2 and σ1 receptors, respectively)
2
Siramesine hydrochloride, MedChemExpress
MedChemExpress Siramesine (Lu 28-179) hydrochloride is a potent sigma-2 receptor agonist. Siramesine hydrochloride has a subnanomolar affinity for sigma-2 receptors (IC50=0.12 nM) and exhibits a 140-fold selectivity for sigma-2 receptors over sigma-1 receptors (IC50=17 nM). Siramesine hydrochloride triggers cell death through destabilisation of mitochondria, but not lysosomes. Anti-cancer activity.