Learn More
Siramesine hydrochloride, MedChemExpress
Código de producto. 16482137
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
5 miligramos
50 miligramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
16482137 50 mg 50 miligramos
16456416 100 mg 100 miligramos
16455998 10 mg 10 miligramos
16438617 5 mg 5 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Código de producto. 16482137 Proveedor Medchem Express N.º de proveedor HY14221A
MedChemExpress Siramesine (Lu 28-179) hydrochloride is a potent sigma-2 receptor agonist. Siramesine hydrochloride has a subnanomolar affinity for sigma-2 receptors (IC50=0.12 nM) and exhibits a 140-fold selectivity for sigma-2 receptors over sigma-1 receptors (IC50=17 nM). Siramesine hydrochloride triggers cell death through destabilisation of mitochondria, but not lysosomes. Anti-cancer activity.
Siramesine (Lu 28-179) hydrochloride is a potent sigma-2 receptor agonist. Siramesine hydrochloride has a subnanomolar affinity for sigma-2 receptors (IC50=0.12 nM) and exhibits a 140-fold selectivity for sigma-2 receptors over sigma-1 receptors (IC50=17 nM). Siramesine hydrochloride triggers cell death through destabilisation of mitochondria, but not lysosomes. Anti-cancer activity.
Especificaciones
Nombre del producto químico o material Siramesine hydrochloride
CAS 224177-60-0
Color Blanco
Notas de grado de pureza Research
Fórmula molecular C30H32ClFN2O
Cantidad 50 mg
Sinónimo Lu 28-179 hydrochloride
Información de solubilidad DMSO : ≥ 42 mg/mL (85.53 mM)
SMILES FC1=CC=C(N2C=C(CCCCN3CCC4(CC3)OCC5=C4C=CC=C5)C6=C2C=CC=C6)C=C1.Cl
Peso molecular (g/mol) 491.04
Formula Weight (peso de la fórmula) 491.04
Porcentaje de pureza 99.05%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Cancer-programmed cell death