Learn More
Tocris Bioscience™ Siramesine Hydrochloride
Descripción
Siramesine hydrochloride is a high affinity σ receptor agonist (Ki values are 0.19 and 17 nM for σ2 and σ1 receptors, respectively). Displays anxiolytic and antidepressant effects in rodents. Also induces cell death in tumor cell lines.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 1'-[4-[1-(4-Fluorophenyl)-1H-indol-3-yl]butyl]spiro[isobenzofuran-1(3H),4'-piperidine hydrochloride |
| CAS | 224177-60-0 |
| Cantidad | 25 mg |
| Fórmula molecular | C30H31FN2O.HCl |
| Formula Weight (peso de la fórmula) | 491.04 |
| Pureza | 98% |