CAS RN 2123491-32-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
CLK1-IN-1, MedChemExpress
MedChemExpress CLK1-IN-1 is a potent and selective of Cdc2-like kinase 1 (CLK1) inhibitor, with an IC50 of 2 nM.