CAS RN 212201-70-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
Ipfencarbazone, MedChemExpress
MedChemExpress Ipfencarbazone is a substance being developed for the control of weeds such as watergrass in rice; herbicide agent.