CAS RN 209984-58-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
LY-411575 isomer 1, MedChemExpress
MedChemExpress LY-411575 isomer 1 is an isomer of LY411575, which is a potent γ-secretase inhibitor.