CAS RN 2080306-23-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
4
de
4
resultados
1
Tocris Bioscience™ BAY 299
Potent and selective BRD1 and TAF1 inhibitor
2
BAY-299, MedChemExpress
MedChemExpress BAY-299 is a very potent, dual inhibitor with IC50s of 67 nM for BRPF2 bromodomains (BD), 8 nM for TAF1 BD2, and 106 nM for TAF1L BD2.
3
4