CAS RN 2035814-50-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
JNJ-61432059, MedChemExpress
MedChemExpress JNJ-61432059 is an oral active and selective negative modulator of AMPAR associated with trans-membrane AMPAR regulatory protein (TARP) γ-8, with a pIC50 of 9.7 for GluA1/γ-8. Exhibits time- and dose-dependent AMPAR/γ-8 receptor occupancy in mouse hippocampus, resulting in robust seizure protection in corneal kindling and pentylenetetrazole (PTZ) anticonvulsant models.