Learn More
JNJ-61432059, MedChemExpress
Código de producto. 16470827
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
5 miligramos
50 miligramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
16470827 50 mg 50 miligramos
16437237 5 mg 5 miligramos
16405386 100 mg 100 miligramos
16404528 10 mg 10 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Código de producto. 16470827 Proveedor Medchem Express N.º de proveedor HY111751
MedChemExpress JNJ-61432059 is an oral active and selective negative modulator of AMPAR associated with trans-membrane AMPAR regulatory protein (TARP) γ-8, with a pIC50 of 9.7 for GluA1/γ-8. Exhibits time- and dose-dependent AMPAR/γ-8 receptor occupancy in mouse hippocampus, resulting in robust seizure protection in corneal kindling and pentylenetetrazole (PTZ) anticonvulsant models.
JNJ-61432059 is an oral active and selective negative modulator of AMPAR associated with trans-membrane AMPAR regulatory protein (TARP) γ-8, with a pIC50 of 9.7 for GluA1/γ-8. Exhibits time- and dose-dependent AMPAR/γ-8 receptor occupancy in mouse hippocampus, resulting in robust seizure protection in corneal kindling and pentylenetetrazole (PTZ) anticonvulsant models.
Especificaciones
Nombre del producto químico o material JNJ-61432059
CAS 2035814-50-5
Color Off-White
Notas de grado de pureza Research
Fórmula molecular C25H22FN5O2
Cantidad 50 mg
Información de solubilidad DMSO : 50 mg/mL (112.75 mM; Need ultrasonic)
SMILES OC1CCN(C2=NC=CC3=C(C4=CC=C(NC(C5)=O)C5=C4)C(C6=CC=C(F)C=C6)=NN32)CC1
Peso molecular (g/mol) 443.47
Formula Weight (peso de la fórmula) 443.47
Porcentaje de pureza 98.75%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Neuroscience-Neuromodulation