CAS RN 200803-37-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
OSIP-486823, MedChemExpress
MedChemExpress OSIP-486823 is a novel microtubule-interfering agent with distinct biological effects on both protein kinase G (PKG) and microtubules.