CAS RN 1891087-61-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
Tocris Bioscience™ BAY 1816032
Potent and selective BUB1 kinase inhibitor
2
BAY-1816032, MedChemExpress
MedChemExpress BAY-1816032 is a potent and oral available BUB1 (budding uninhibited by benzimidazoles 1) kinase inhibitor with an IC50 of 7 nM.