CAS RN 188968-51-6
IUPAC Nombre:
2-[(2S,5R,8S,11S)-5-benzyl-11-{3-[(diaminomethylidene)amino]propyl}-7-methyl-3,6,9,12,15-pentaoxo-8-(propan-2-yl)-1,4,7,10,13-pentaazacyclopentadecan-2-yl]acetic acid
Sinónimos:
cilengitide
Peso molecular (g/mol):
588.67
Fórmula molecular:
C27H40N8O7
InChi Key:
AMLYAMJWYAIXIA-VWNVYAMZSA-N
SMILES:
CC(C)[C@@H]1N(C)C(=O)[C@@H](CC2=CC=CC=C2)NC(=O)[C@H](CC(O)=O)NC(=O)CNC(=O)[C@H](CCCN=C(N)N)NC1=O
1
–
2
de
2
resultados
1
Tocris Bioscience™ Cilengitide
Potent and selective inhibitor of integrins αvβ3 and αvβ5
2
Cilengitide, MedChemExpress
MedChemExpress Cilengitide (EMD 121974) is a potent and selective inhibitor of the integrins ανβ3 and ανβ5. Cilengitide inhibits binding of isolated ανβ3 and ανβ5 to Vitronectin with an IC50 value of 4 and 79 nM, respectively .