CAS RN 188343-77-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
MBP146-78, MedChemExpress
MedChemExpress MBP146-78 is a potent and selective inhibitor of cGMP dependent protein kinases.