CAS RN 186543-64-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
LY 344864 racemate, MedChemExpress
MedChemExpress LY 344864 racemate is a 5-HT1F receptor agonist extracted from patent US 5708187 A.