CAS RN 1820708-73-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
SKA-121, MedChemExpress
MedChemExpress SKA-121 is a selective KCa3.1 activator. SKA-121 exhibits EC50s of 109 nM and 4.4 μM for KCa3.1 and KCa2.3, respectively.
2
Tocris™ SKA 121
Positive allosteric modulator of KCa3.1 (IK) channels