Learn More
Tocris™ SKA 121
Descripción
SKA 121 is a positive allosteric modulator of KCa3.1 (intermediate-conductance Ca2+-activated potassium) channels (EC50 = 109 nM). Displays 40-fold selectivity for KCa3.1 over KCa2.3, and 200 to 400-fold selectivity over KV and NaV channels. Potentiates bradykinin-induced vasodilation ex vivo. Lowers mean arterial blood pressure in normotensive and hypertensive mice.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 5-Methylnaphth[2,1-d]oxazol-2-amine |
| CAS | 1820708-73-3 |
| Cantidad | 5 mg |
| Objetivo | Ca2+-Activated Potassium Channel Activators |
| Fórmula molecular | C12H10N2O |
| Formula Weight (peso de la fórmula) | 960 |
| Pureza | 98% |