CAS RN 1798871-31-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
resultados
1
Tocris Bioscience™ Centrinone B
High affinity and selective PLK4 inhibitor
2
Centrinone-B, MedChemExpress
MedChemExpress Centrinone-B (LCR-323) is a potent and highly selective PLK4 inhibitor, with a Ki of 0.59 nM.
3