Learn More
Tocris Bioscience™ Centrinone B
Descripción
Centrinone B is a high affinity and selective PLK4 inhibitor (Ki = 0.6 nM). Exhibits >2000-fold selectivity for PLK4 over Aurora A and Aurora B. Depletes centriole and centrosome levels in vitro. Induces cell cycle arrest in normal human cell lines in a p53-dependent manner.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 2-[[2-Fluoro-4-[[(2-fluoro-3-nitrophenyl)methyl]sulfonyl]phenyl]thio]-5-methoxy-N-(5-methyl-1H-pyrazol-3-yl)-6-(1-piperidinyl)-4-pyrimidinamine |
| CAS | 1798871-31-4 |
| Sinónimo | LCR 323 |
| Cantidad | 10 mg |
| Objetivo | Polo-like Kinase Inhibitors |
| Fórmula molecular | C27H27F2N7O5S2 |
| Formula Weight (peso de la fórmula) | 631.67 |
| Pureza | >98% |