CAS RN 1672691-74-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
Tocris Bioscience™ JZP 430
Potent and irreversible α/β-hydrolase domain 6 (ABHD6) inhibitor
2
JZP-430, MedChemExpress
MedChemExpress JZP-430 is a potent, highly selective, irreversible inhibitor of α/β-hydrolase domain 6 (ABHD6) with an IC50 of 44 nM, exhibits ∼230-fold selectivity over fatty acid amide hydrolase (FAAH) and lysosomal acid lipase (LAL).