CAS RN 164204-38-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
Tocris Bioscience™ Phosphoramidon disodium salt
Neutral endopeptidase and endothelin converting enzyme inhibitor
2
Phosphoramidon Disodium, MedChemExpress
MedChemExpress Phosphoramidon Disodium is a metalloprotease inhibitor. Phosphoramidon inhibits endothelin-converting enzyme (ECE), neutral endopeptidase (NEP), and angiotensin-converting enzyme (ACE) with IC50 values of 3.5, 0.034, and 78 μM, respectively.