CAS RN 1639417-54-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Tenalisib R Enantiomer, MedChemExpress
MedChemExpress Tenalisib R Enantiomer (RP6530 R Enantiomer) is an R enantiomer of Tenalisib. Tenalisib is a potent and selective PI3Kδ and PI3Kγ inhibitor with IC50 values of 25 and 33 nM, respectively.