CAS RN 1608125-21-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
AMG319, MedChemExpress
MedChemExpress AMG319 is a potent and selective PI3Kδ kinase inhibitor with IC50 of 18 nM.