CAS RN 157115-85-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
1-(2-Fenilacetil)-L-prolinglicina Éster de Etilo, TRC
CAS: 157115-85-0 Fórmula molecular: C17 H22 N2 O4 Peso molecular (g/mol): 318.37 Sinónimo: Glycine, 1-(2-phenylacetyl)-L-prolyl-, ethyl ester,Glycine, 1-(phenylacetyl)-L-prolyl-, ethyl ester (9CI),Glycine, N-[1-(phenylacetyl)-L-prolyl]-, ethyl ester,1-(2-Phenylacetyl)-L-prolylglycine ethyl ester,GVS 111,Noopept,Omberacetam,SGS 111 Nombre IUPAC: Etilacetato 2-[[(2S)-1-(2-fenilacetil)pirrolidina-2-carbonil]amino]acetato SMILES: CCOC(=O)CNC(=O)[C@@H]1CCCN1C(=O)Cc2ccccc2
2
Omberacetam, MedChemExpress
MedChemExpress Omberacetam (GVS-111) is a medication promoted and prescribed in Russia and neighbouring countries as a nootropic.