CAS RN 153587-01-0
IUPAC Nombre:
1-[2-hydroxy-5-(trifluoromethyl)phenyl]-5-(trifluoromethyl)-2,3-dihydro-1H-1,3-benzodiazol-2-one
Sinónimos:
1-[2-hydroxy-5-(trifluoromethyl)phenyl]-5-(trifluoromethyl)-3H-1,3-benzodiazol-2-one
Peso molecular (g/mol):
362.23
Fórmula molecular:
C15H8F6N2O2
InChi Key:
YLFMCMWKHSDUCT-UHFFFAOYSA-N
SMILES:
OC1=CC=C(C=C1N1C(=O)NC2=CC(=CC=C12)C(F)(F)F)C(F)(F)F
1
–
2
de
2
resultados
2
NS-1619, MedChemExpress
MedChemExpress NS-1619 is an opener of large conductance Ca2+-activated K+ (BK) channel. NS-1619 is a highly effective relaxant with an EC50 of about 10 - 30 μM in several smooth muscles of blood vessels and other tissues. NS1619 inhibits proliferation and induces apoptosis in A2780 ovarian cancer cells.