CAS RN 1532533-69-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Umbralisib R-enantiomer, MedChemExpress
MedChemExpress Umbralisib R-enantiomer (TGR-1202 R-enantiomer) is a PI3Kδ inhibitor, which is the less active enantiomer of TGR-1202.