CAS RN 1527503-11-2
IUPAC Nombre:
5'-methoxy-6'-[3-(pyrrolidin-1-yl)propoxy]spiro[cyclobutane-1,3'-indol]-2'-amine
Sinónimos:
5'-methoxy-6'-[3-(pyrrolidin-1-yl)propoxy]spiro[cyclobutane-1,3'-indol]-2'-amine
Peso molecular (g/mol):
329.44
Fórmula molecular:
C19H27N3O2
InChi Key:
BKCDJTRMYWSXMC-UHFFFAOYSA-N
SMILES:
COC1=C(OCCCN2CCCC2)C=C2N=C(N)C3(CCC3)C2=C1
1
–
2
de
2
resultados
2
A-366, MedChemExpress
MedChemExpress A-366 is a potent, highly selective, peptide-competitive histone methyltransferase G9a inhibitor with IC50s of 3.3 and 38 nM for G9a and GLP (EHMT1), respectively.