CAS RN 1446502-11-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
Enasidenib, TRC
CAS: 1446502-11-9 Fórmula molecular: C19H17F6N7O Peso molecular (g/mol): 473.37 Sinónimo: 2-Methyl-1-[[4-[6-(trifluoromethyl)-2-pyridinyl]-6-[[2-(trifluoromethyl)-4-pyridinyl]amino]-1,3,5-triazin-2-yl]amino]-2-propanol,2-Methyl-1-(4-(6-(trifluoromethyl)pyridin-2-yl)-6-(2-(trifluoromethyl)pyridin-4-ylamino)-1,3,5-triazin-2-ylamino)propan-2-ol,AG 221,CC 90007 Free Base,Enasidenib,Idhifa SMILES: CC(C)(O)CNc1nc(Nc2ccnc(c2)C(F)(F)F)nc(n1)c3cccc(n3)C(F)(F)F
2
Enasidenib, MedChemExpress
MedChemExpress Enasidenib is an oral, potent, reversible, selective inhibitor of the IDH2 mutant enzymes, with IC50s of 100 and 400 nM against IDH2R140Q and IDH2R172K, respectively.