CAS RN 1391054-02-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
Clorhidrato de rac-Vigabatrina, TRC
CAS: 1391054-02-6 Fórmula molecular: C6H12ClNO2 Peso molecular (g/mol): 165.62 Nombre IUPAC: 4-aminohex-5-ácido enoico; Cloruro SMILES: Cl.NC(CCC(=O)O)C=C
2
Vigabatrin hydrochloride, MedChemExpress
MedChemExpress Vigabatrin hydrochloride (γ-Vinyl-GABA hydrochloride), a inhibitory neurotransmitter GABA vinyl-derivative, is an orally active and irreversible GABA transaminase inhibitor. Vigabatrin hydrochloride is an antiepileptic agent, which acts by increasing GABA levels in the brain by inhibiting the catabolism of GABA by GABA transaminase.