CAS RN 1380548-02-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
4
de
4
resultados
1
Tocris Bioscience™ SR 10067
Selective Rev-Erbα/β agonist; anxiolytic
2
SR10067, MedChemExpress
MedChemExpress SR10067 is a potent, selective and brain penetrant Rev-Erbα/β agonist, with IC50 values are 160 and 170 nM for Rev-Erbβ and Rev-Erbα, respectively. SR10067 has anxiolytic activity.
3
4