Learn More
Tocris Bioscience™ SR 10067
Descripción
SR 10067 is a selective Rev-Erbα/β agonist (IC50 values are 160 and 170 nM for Rev-Erbβ and Rev-Erbα respectively). Exhibits no significant activity against a wide range of nuclear receptors, other receptors, ion channels and transporters at 20 μM. Increases wakefulness, reduces REM and slow wave sleep in vivo. Also reduces anxiety-like behavior in marble-burying assay. Cell permeable and brain penetrant.
Especificaciones
Especificaciones
| Nombre del producto químico o material | [3-[[4-(1,1-Dimethylethoxy)phenoxy]methyl]-3,4-dihydro-2(1H)-isoquinolinyl]-1-naphthalenylmethanone |
| CAS | 1380548-02-6 |
| Intervalo de porcentaje de ensayo | >98% |
| Cantidad | 10 mg |
| Fórmula molecular | C31H31NO3 |
| Formula Weight (peso de la fórmula) | Observed MW: 465.58 |