CAS RN 1375465-91-0
IUPAC Nombre:
N-hydroxy-2-[(1-phenylcyclopropyl)amino]pyrimidine-5-carboxamide
Sinónimos:
N-hydroxy-2-[(1-phenylcyclopropyl)amino]pyrimidine-5-carboxamide
Peso molecular (g/mol):
270.29
Fórmula molecular:
C14H14N4O2
InChi Key:
LIIWIMDSZVNYHY-UHFFFAOYSA-N
SMILES:
ONC(=O)C1=CN=C(NC2(CC2)C2=CC=CC=C2)N=C1
1
–
2
de
2
resultados
2
ACY-738, MedChemExpress
MedChemExpress ACY-738 is a potent, selective and orally-bioavailable HDAC6 inhibitor, with an IC50 of 1.7 nM; ACY-738 also inhibits HDAC1, HDAC2, and HDAC3, with IC50s of 94, 128, and 218 nM.