CAS RN 1361030-48-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
LDC1267, MedChemExpress
MedChemExpress LDC1267 is a highly selective TAM (Tyro3, Axl and Mer) kinase inhibitor with IC50s of [1].