CAS RN 1333207-63-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
resultados
1
Tocris Bioscience™ SAR 7334
Potent TRPC6 inhibitor
2
SAR7334 hydrochloride, MedChemExpress
MedChemExpress SAR7334 hydrochloride is a potent and specific TRPC6 inhibitor, inhibiting TRPC6 currents with IC50 of 7.9 nM.
3