Learn More
Tocris Bioscience™ SAR 7334
Descripción
SAR 7334 is a potent TRPC6 inhibitor (IC50 values are 9.5, 226 and 282 nM for TRPC6, TRPC7 and TRPC3-mediated Ca2+ influx). Exhibits no significant activity at TRPC4 and TRPC5 channels. Suppresses TRPC6-dependent acute hypoxic pulmonary vasoconstriction (HPV) in the isolated perfused mouse lung. Orally bioavailable.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 4-[[(1R,2R)-2-[(3R)-3-Amino-1-piperidinyl]-2,3-dihydro-1H-inden-1-yl]oxy]-3-chlorobenzonitrile dihydrochloride |
| CAS | 1333207-63-8 |
| Intervalo de porcentaje de ensayo | >98% |
| Cantidad | 5 mg |
| Fórmula molecular | C21H22ClN3O.2HCl |
| Formula Weight (peso de la fórmula) | Observed MW: 440.79 |