CAS RN 1313613-09-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
Tocris Bioscience™ AMZ 30
Potent, selective and irreversible protein phosphatase methylesterase-1 (PME-1) inhibitor
2
AMZ30, MedChemExpress
MedChemExpress AMZ30 is a selective, covalent inhibitors of protein phosphatase methylesterase-1(PME-1; IC50=600 nM); selectively inactivates PME-1 and reduces the demethylated form of PP2A in living cells.