CAS RN 1242156-23-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
RN486, MedChemExpress
MedChemExpress RN486 is a potent, selective and orally active Btk inhibitor with an IC50 of 4.0 nM and a Kd of 0.31 nM. RN486 is less active for other kinases. RN486 can be used for rheumatoid arthritis and systemic lupus erythematosus research.