CAS RN 1221277-90-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
resultados
1
Tocris Bioscience™ GSK 2033
Potent LXR antagonist
2
GSK2033, MedChemExpress
MedChemExpress GSK2033 is a LXR antagonist with pIC50s of 7 and 7.4 for LXRα or LXRβ, respectively.
3