CAS RN 1220646-23-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
AVN-492, MedChemExpress
MedChemExpress AVN-492 is a very specific and highly-selective antagonist with picomolar affinity to 5-HT6R (Ki=91 pM).