CAS RN 1218777-13-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
CAY10505, MedChemExpress
MedChemExpress CAY10505 is a potent and selective PI3Kγ inhibitor with an IC50 of 30 nM in neurons.