CAS RN 1198117-23-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Sodium Channel inhibitor 1, MedChemExpress
MedChemExpress Sodium Channel inhibitor1, one of 3-Oxoisoindoline-1-carboxamides, is a novel and selective voltage-gated sodium channel for pain treatment.