CAS RN 1139889-93-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Golgicide A, MedChemExpress
MedChemExpress Golgicide A (GCA) is a potent, highly specific, and reversible inhibitor of the cis-Golgi ADP-ribosylation factor guanine nucleotide exchange factors (ArfGEF) GBF1. Golgicide A drastically reduced replication of coxsackievirus B3 (CVB3) and other human enterovirus species.