missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
ZNF385D Polyclonal antibody specifically detects ZNF385D in Human,Mouse samples. It is validated for ELISA,Western Blot
Especificaciones
Especificaciones
| Antígeno | ZNF385D |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Polyclonal |
| Conjugado | DyLight 680 |
| Formulación | 50mM Sodium Borate |
| Alias de gen | FLJ12586, zinc finger protein 329 |
| Especie del huésped | Rabbit |
| Inmunógeno | zinc finger protein 385D. (Uniprot# Q9H6B1) |
| Método de purificación | Immunogen affinity purified |
| Cantidad | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?