Learn More
Isofield™ Zetta 230 Non-Sterile TieOon Cleanroom Facemask - ISO Class 4/5 Compatible
Click to view available options
:
Tamaño: Ancha
Descripción
- Non-sterile tie-on cleanroom facemask
- 230 mm length with malleable nose strip
- 3-layer non-woven configuration
- Highly efficient bacterial and particle filtration
- Particle Shedding (Helmke Drum) Category I
- Individually packed to maintain low CFU count
- Pouches printed with IPA resistant ink to maintain traceability
Especificaciones
Especificaciones
| Tipo de producto | Cleanroom Face Mask |
| Color | Blanco |
| Fastening Type | Lazos |
| Material | Polyethylene, Polypropylene |
| Tamaño | Wide |
| Esterilidad | No estéril |
| Modelo | Plisado |